Assay Details
Gene Symbol: AOPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627644.1 | XP_023483412.1 | 2494 | 83 | ||
XM_023627648.1 | XP_023483416.1 | 2495 | 83 | |||
XM_023627650.1 | XP_023483418.1 | 2196 | 83 | |||
XM_023627651.1 | XP_023483419.1 | 2494 | 83 | |||
XM_023627652.1 | XP_023483420.1 | 2494 | 83 | |||
XM_023627653.1 | XP_023483421.1 | 2494 | 83 | |||
XM_023627654.1 | XP_023483422.1 | 2494 | 83 | |||
XM_023627655.1 | XP_023483423.1 | 2494 | 83 | |||
XM_023627659.1 | XP_023483427.1 | 2494 | 83 | |||
XM_023627661.1 | XP_023483429.1 | 2197 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
