Assay Details
Gene Symbol: TUT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627112.1 | XP_023482880.1 | 1360 | 63 | ||
XM_023627113.1 | XP_023482881.1 | 1360 | 63 | |||
GenBank mRNA | JL640534.1 | - | 8 - 9 | 1275 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
