Assay Details
Gene Symbol: PSAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496362.3 | XP_001496412.2 | 498 | 74 | ||
XM_023627166.1 | XP_023482934.1 | 432 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
