Assay Details
Gene Symbol: GNAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627175.1 | XP_023482943.1 | 450 | 74 | ||
XM_023627176.1 | XP_023482944.1 | 415 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
