Assay Details
Gene Symbol: GNA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627177.1 | XP_023482945.1 | 338 | 88 | ||
XM_023627178.1 | XP_023482946.1 | 243 | 88 | |||
XM_023627179.1 | XP_023482947.1 | 338 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
