Assay Details
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491923.5 | XP_001491973.1 | 2893 | 59 | ||
XM_005604984.3 | XP_005605041.1 | 2407 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
