Assay Details
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627642.1 | XP_023483410.1 | 1574 | 61 | ||
XM_023627643.1 | XP_023483411.1 | 1437 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
