Assay Details
Gene Symbol: ERO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627735.1 | XP_023483503.1 | 924 | 66 | ||
XM_023627736.1 | XP_023483504.1 | 900 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
