Assay Details
Gene Symbol: GCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627771.1 | XP_023483539.1 | 665 | 93 | ||
XM_023627773.1 | XP_023483541.1 | 666 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
