Assay Details
Gene Symbol: L3HYPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735623.2 | XP_014591109.2 | 886 | 70 | ||
XM_023627824.1 | XP_023483592.1 | 890 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
