Assay Details
Gene Symbol: PIGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499883.5 | XP_001499933.1 | 1601 | 85 | ||
XM_005605263.3 | XP_005605320.1 | 1569 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
