Assay Details
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005605326.3 | XP_005605383.2 | 158 | 72 | ||
XM_005605327.3 | XP_005605384.2 | 158 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
