Assay Details
Gene Symbol: CIPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916581.4 | XP_001916616.1 | 515 | 99 | ||
XM_014735739.2 | XP_014591225.1 | 445 | 99 | |||
XM_023628161.1 | XP_023483929.1 | 513 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
