Assay Details
Gene Symbol: PPP4R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628282.1 | XP_023484050.1 | 1287 | 65 | ||
XM_023628283.1 | XP_023484051.1 | 1287 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
