Assay Details
Gene Symbol: CATSPERB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628284.1 | XP_023484052.1 | 2671 | 59 | ||
XM_023628285.1 | XP_023484053.1 | 2575 | 59 | |||
XM_023628286.1 | XP_023484054.1 | 2519 | 59 | |||
XM_023628287.1 | XP_023484055.1 | 2380 | 59 | |||
XM_023628288.1 | XP_023484056.1 | 2356 | 59 | |||
XM_023628289.1 | XP_023484057.1 | 2364 | 59 | |||
XM_023628290.1 | XP_023484058.1 | 2262 | 59 | |||
XM_023628291.1 | XP_023484059.1 | 2066 | 59 | |||
XM_023628292.1 | XP_023484060.1 | 2171 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
