Assay Details
Gene Symbol: WARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488721.6 | XP_001488771.3 | 554 | 68 | ||
XM_023628399.1 | XP_023484167.1 | 443 | 68 | |||
XM_023628400.1 | XP_023484168.1 | 476 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
