Assay Details
Gene Symbol: RCOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628439.1 | XP_023484207.1 | 1411 | 65 | ||
XM_023628440.1 | XP_023484208.1 | 1249 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
