Assay Details
Gene Symbol: NUDT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005605565.3 | XP_005605622.2 | 430 | 58 | ||
XM_023628582.1 | XP_023484350.1 | 283 | 58 | |||
XM_023628583.1 | XP_023484351.1 | 520 | 58 | |||
XM_023628584.1 | XP_023484352.1 | 552 | 58 | |||
XM_023628586.1 | XP_023484354.1 | 335 | 58 | |||
XM_023628587.1 | XP_023484355.1 | 209 | 58 | |||
XM_023628588.1 | XP_023484356.1 | 591 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
