Assay Details
Gene Symbol: DCAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629691.1 | XP_023485459.1 | 1376 | 77 | ||
XM_023629692.1 | XP_023485460.1 | 1187 | 77 | |||
XM_023629693.1 | XP_023485461.1 | 1376 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
