Assay Details
Gene Symbol: ELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735945.2 | XP_014591431.2 | 521 | 58 | ||
XM_023629846.1 | XP_023485614.1 | 706 | 58 | |||
XM_023629847.1 | XP_023485615.1 | 706 | 58 | |||
XM_023629848.1 | XP_023485616.1 | 852 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
