Assay Details
Gene Symbol: ECPAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628638.1 | XP_023484406.1 | 2486 | 64 | ||
XM_023628639.1 | XP_023484407.1 | 2693 | 64 | |||
XM_023628640.1 | XP_023484408.1 | 2757 | 64 | |||
XM_023628641.1 | XP_023484409.1 | 2586 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
