Assay Details
Gene Symbol: PTGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628647.1 | XP_023484415.1 | 419 | 67 | ||
XM_023628648.1 | XP_023484416.1 | 418 | 67 | |||
XM_023628649.1 | XP_023484417.1 | 260 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
