Assay Details
Gene Symbol: MEGF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005605745.3 | XP_005605802.2 | 925 | 62 | ||
XM_023628768.1 | XP_023484536.1 | 932 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
