Assay Details
Gene Symbol: TRAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014736011.2 | XP_014591497.1 | 2901 | 58 | ||
XM_023628773.1 | XP_023484541.1 | 2978 | 58 | |||
XM_023628774.1 | XP_023484542.1 | 2372 | 58 | |||
XM_023628775.1 | XP_023484543.1 | 2375 | 58 | |||
XM_023628776.1 | XP_023484544.1 | 2202 | 58 | |||
XM_023628777.1 | XP_023484545.1 | 2268 | 58 | |||
XM_023628779.1 | XP_023484547.1 | 2808 | 58 | |||
XM_023628780.1 | XP_023484548.1 | 2512 | 58 | |||
XM_023628781.1 | XP_023484549.1 | 2032 | 58 | |||
XM_023628782.1 | XP_023484550.1 | 2098 | 58 | |||
XM_023628783.1 | XP_023484551.1 | 1736 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
