Assay Details
Gene Symbol: PSMB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502120.5 | XP_001502170.1 | 150 | 63 | ||
GenBank mRNA | HQ889915.1 | - | 1 - 2 | 83 | 63 | |
JL616288.1 | - | 1 - 2 | 78 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
