Assay Details
Gene Symbol: TOR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501467.5 | XP_001501517.2 | 602 | 56 | ||
XM_003364143.4 | XP_003364191.2 | 602 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
