Assay Details
Gene Symbol: PTGES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628980.1 | XP_023484748.1 | 1010 | 72 | ||
XM_023628981.1 | XP_023484749.1 | 1095 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
