Assay Details
Gene Symbol: SPOUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499831.6 | XP_001499881.3 | 104 | 64 | ||
XM_005605852.3 | XP_005605909.1 | 104 | 64 | |||
XM_023629069.1 | XP_023484837.1 | 104 | 64 |

More Information
Set Membership: |
![]() ![]() |
