Assay Details
Gene Symbol: SPOUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499831.6 | XP_001499881.3 | 890 | 65 | ||
XM_005605852.3 | XP_005605909.1 | 686 | 65 | |||
XM_023629069.1 | XP_023484837.1 | 959 | 65 | |||
XM_023629070.1 | XP_023484838.1 | 1141 | 65 | |||
XM_023629071.1 | XP_023484839.1 | 1017 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
