Assay Details
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504498.5 | XP_001504548.1 | 935 | 98 | ||
XM_005611220.3 | XP_005611277.1 | 846 | 98 | |||
XM_023643642.1 | XP_023499410.1 | 695 | 98 | |||
XM_023643643.1 | XP_023499411.1 | 512 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
