Assay Details
Gene Symbol: NABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504827.4 | XP_001504877.1 | 414 | 72 | ||
XM_005611303.3 | XP_005611360.1 | 236 | 72 | |||
XM_005611304.3 | XP_005611361.1 | 367 | 72 | |||
XM_023643786.1 | XP_023499554.1 | 419 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
