Assay Details
Gene Symbol: UCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499306.5 | XP_001499356.2 | 478 | 71 | ||
XM_014736098.2 | XP_014591584.1 | 483 | 71 | |||
XM_023629187.1 | XP_023484955.1 | 488 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
