Assay Details
Gene Symbol: GTF3C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629209.1 | XP_023484977.1 | 511 | 65 | ||
XM_023629210.1 | XP_023484978.1 | 511 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
