Assay Details
Gene Symbol: GFI1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364167.3 | XP_003364215.2 | 683 | 59 | ||
XM_014736113.2 | XP_014591599.1 | 694 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
