Assay Details
Gene Symbol: CCDC187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023629344.1 | XP_023485112.1 | 3900 | 71 | ||
XM_023629345.1 | XP_023485113.1 | 2834 | 71 | |||
XM_023629346.1 | XP_023485114.1 | 2915 | 71 | |||
XM_023629347.1 | XP_023485115.1 | 3398 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
