Assay Details
Gene Symbol: SAPCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917502.4 | XP_001917537.2 | 1950 | 76 | ||
XM_023629429.1 | XP_023485197.1 | 2021 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
