Assay Details
Gene Symbol: FAM166A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491293.4 | XP_001491343.1 | 923 | 68 | ||
XM_005605964.3 | XP_005606021.1 | 923 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
