Assay Details
Gene Symbol: N6AMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496200.6 | XP_001496250.4 | 187 | 93 | ||
XM_005606113.3 | XP_005606170.2 | 187 | 93 | |||
XM_023629979.1 | XP_023485747.1 | 187 | 93 | |||
XM_023629980.1 | XP_023485748.1 | 187 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
