Assay Details
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014736328.2 | XP_014591814.1 | 719 | 70 | ||
XM_014736329.2 | XP_014591815.1 | 217 | 70 | |||
XM_014736330.2 | XP_014591816.1 | 516 | 70 | |||
XM_023629985.1 | XP_023485753.1 | 1167 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
