Assay Details
Gene Symbol: CLIC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493952.4 | XP_001494002.2 | 1540 | 78 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CLIC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493952.4 | XP_001494002.2 | 1540 | 78 |