Assay Details
Gene Symbol: IGSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014736382.2 | XP_014591868.1 | 1590 | 57 | ||
XM_014736383.2 | XP_014591869.1 | 1339 | 57 | |||
XM_014736384.2 | XP_014591870.1 | 1211 | 57 | |||
XM_023630194.1 | XP_023485962.1 | 1232 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
