Assay Details
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488579.3 | XP_001488629.1 | 1516 | 88 | ||
XM_014736518.2 | XP_014592004.1 | 1322 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
