Assay Details
Gene Symbol: GPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023630587.1 | XP_023486355.1 | 1494 | 62 | ||
XM_023630588.1 | XP_023486356.1 | 1932 | 62 | |||
XM_023630589.1 | XP_023486357.1 | 1458 | 62 | |||
XM_023630590.1 | XP_023486358.1 | 1497 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
