Assay Details
Gene Symbol: PLPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491885.6 | XP_001491935.3 | 353 | 98 | ||
XM_005606317.3 | XP_005606374.2 | 353 | 98 | |||
XM_005606318.3 | XP_005606375.2 | 353 | 98 | |||
XM_005606319.3 | XP_005606376.2 | 669 | 98 | |||
XM_005606320.3 | XP_005606377.2 | 353 | 98 | |||
XM_005606321.3 | XP_005606378.1 | 452 | 98 | |||
XM_014736568.2 | XP_014592054.2 | 353 | 98 | |||
XM_023630618.1 | XP_023486386.1 | 353 | 98 | |||
XM_023630619.1 | XP_023486387.1 | 261 | 98 | |||
XM_023630621.1 | XP_023486389.1 | 332 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023630621.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
