Assay Details
Gene Symbol: BAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493179.3 | XP_001493229.1 | 327 | 80 | ||
XM_023630616.1 | XP_023486384.1 | 328 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
