Assay Details
Gene Symbol: GOT1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492126.6 | XP_001492176.4 | 437 | 65 | ||
XM_023630740.1 | XP_023486508.1 | 437 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
