Assay Details
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915437.5 | XP_001915472.3 | 820 | 66 | ||
XM_023630694.1 | XP_023486462.1 | 820 | 66 | |||
XM_023630695.1 | XP_023486463.1 | 560 | 66 | |||
XM_023630696.1 | XP_023486464.1 | 592 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
