Assay Details
Gene Symbol: DUSP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494228.5 | XP_001494278.1 | 309 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001494228.5 transcripts.
Set Membership: |
![]() ![]() |
