Assay Details
Gene Symbol: DUSP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494228.5 | XP_001494278.1 | 820 | 75 | ||
XM_005606332.3 | XP_005606389.1 | 874 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
