Assay Details
Gene Symbol: FUT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493209.4 | XP_001493259.1 | 1421 | 64 | ||
XM_014736576.2 | XP_014592062.1 | 936 | 64 | |||
XM_023630855.1 | XP_023486623.1 | 849 | 64 | |||
XM_023630856.1 | XP_023486624.1 | 936 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
